Order Entry
Belgium
ContactUsLinkComponent
Water, Tracepur® for inorganic trace analysis, Supelco®
Water, Tracepur® for inorganic trace analysis, Supelco®
Catalog # 1.00473.5000
Supplier:  Merck
CAS Number:  
Water, Tracepur® for inorganic trace analysis, Supelco®
Catalog # 1.00473.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Molar Mass:
    18.02 g/mol
  • Boiling Point:
    100 °C (1013 hPa)
  • Vapor Pressure:
    23 hPa (20 °C)
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332