Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water

Supplier:   Bernd Kraft
Water
Supplier:  Bernd Kraft
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 110,00 €
    110,00 €
    1 * 2.500 mL
       110,00 €

  • Plastic bottle
    0,5 L
    Log in for availability
    Log in for availability
    1 * 0,5 L
    1 * 0,5 L

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 63,10 €
    63,10 €
    1 * 1 L
       63,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.