Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Supplier:   MARIENFELD
Staining jars
Supplier:  MARIENFELD
Certificates
Staining jars
4 Options Available Below
Product Details & Documents
Staining set consists of glass dish and staining jar made of soda lime glass, and staining tray made of either pressed glass or stainless steel.
These staining tray hold up to 10 slides (76×26 mm).
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • MARI6611000
    Wire handle for staining tray
    Wire handle for staining tray
    - mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 35,40 €
    35,40 €
    1 * 10 ST
       35,40 €
  • MARI4200004
    Staining jar, with lid
    Staining jar, with lid
    105×85×70 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 288,00 €
    288,00 €
    1 * 10 ST
       288,00 €
  • MARI6618000
    Staining tray, stainless steel
    Staining tray, stainless steel
    - mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 287,00 €
    287,00 €
    1 * 10 ST
       287,00 €
  • MARI4200005
    Staining tray, pressed glass
    Staining tray, pressed glass
    - mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 198,00 €
    198,00 €
    1 * 10 ST
       198,00 €