ContactUsLinkComponent
Staining jars acc. to Schiefferdecker
Staining jars acc. to Schiefferdecker
Catalog # 631-0699
Supplier:  MARIENFELD
undefined
Staining jars acc. to Schiefferdecker
Catalog # 631-0699
Supplier:  MARIENFELD
Specifications
  • Description:
    Staining jar, Schiefferdecker
  • W×D×H:
    89×70,5×46 mm
  • Pk:
    10
Specifications
About this item
Made of soda-lime glass, with cover.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.