Water ≥99%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Catalog # 34478-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Non-volatile matter:Max. 0.0005 %
- Suitability for residue analysis:Max. 5 ng/l
- Determination of P/N:≤5 ng/L
Specification Test Results
About this item
:
:18.02 g/mol
Merck Index:12,10175
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332