Orders LinkContactUsLinkComponent
Douche cans/irrigators
Douche cans/irrigators
2 l, Douche can with flat back and suspension tag
Catalog # KART590
Supplier:   KARTELL
Douche cans/irrigators
Catalog # KART590
Supplier:  KARTELL
Douche cans/irrigators
Douche cans/irrigators
2 l, Douche can with flat back and suspension tag
Price
16,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Douche cans/irrigators, made of polypropylene.
  • Translucent
  • With flat back and suspension tag
  • Autoclavable
Specifications
  • Type:
    Douche can with flat back and suspension tag
  • Capacity:
    2 l
  • Seq:
    02