Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water, extra pure, deionised

Water, extra pure, deionised

Water, extra pure, deionised
Certificates
Water, extra pure, deionised

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (760 mmHg)
  • Melting Pt:0 °C
  • Density:1 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Residue after evaporation:
    ≤25 ppm
  • Conductivity:
    ≤5.0 µS/cm
Product Family Options

Product Information