Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium nitrate ≥99.994% (metals basis)

Potassium nitrate ≥99.994% (metals basis)

Potassium nitrate ≥99.994% (metals basis)
Certificates
Potassium nitrate ≥99.994% (metals basis)
Potassium nitrate ≥99.994% (metals basis)

2 Options Available Below

Product Details & Documents

Purity: 99.994% CAS No.: 7757-79-1 Molecular Formula: KNO3 Formula Weight: 101.11 Physical Form: Crystalline Density: 2.109 Appearance: White Odour: Odourless Melting Point: 334° Boiling Point: 400° dec. Merck Reference: 14,7648 UN No.: 1486 MDL No.: MFCD00011409

Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:14,7648
  • Boiling Pt:400 °C (dec.)
  • Melting Pt:334 °C
  • Density:2.109
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.