Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium iodide

Potassium iodide

Glass bottle for solids, 500 g
Catalog # WAKO168-03975
Marketsource
Supplier:   Wako
Potassium iodide
Catalog # WAKO168-03975
Supplier:  Wako
Marketsource
CAS Number:  7681-11-0
Potassium iodide

Glass bottle for solids, 500 g

CAS Number:  7681-11-0
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (1013 hPa)
  • Melting Pt:686 °C
  • Density:3.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.