Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium iodide ≥99%, GPR RECTAPUR®

Potassium iodide ≥99%, GPR RECTAPUR®

Supplier:   VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Potassium iodide ≥99%, GPR RECTAPUR®

5 Options Available Below

Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (1013 hPa)
  • Melting Pt:681 °C
  • Density:3.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 0.1 %
  • Fe (Iron):
    Max. 10 ppm
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.