Order Entry
Belgium
ContactUsLinkComponent
Potassium iodide ≥99.5%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Health
Potassium iodide ≥99.5%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Health
Potassium iodide ≥99.5%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1430 g/l (20 °C)
  • Melting Point:
    723 °C
  • Molar Mass:
    166.00 g/mol
  • Bulk Density:
    1500 kg/m3
  • Boiling Point:
    1325 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (745 °C)
  • Density:
    3.13 g/cm3 (20 °C)
  • pH:
    6.9 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together