Order Entry
Belgium
ContactUsLinkComponent
Potassium iodide AST, 1 g, tablets
Health
Potassium iodide AST, 1 g, tablets
Catalog # 296322E
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Potassium iodide AST, 1 g, tablets
Catalog # 296322E
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 Tab.
  • Seq:
    0940

Specifications

Specification Test Results

  • Appearance:
    White biconvex tablets
  • Weight uniformity::
    -
  • Max. 10 % outside 950–1050 mg:
    Conforms
  • None outside 930–1070 mg:
    Conforms
  • Average tablet weight:
    980 --> 1020 mg
  • Diameter:
    11.0 --> 11.5 mm
  • Hardness (kp) (off machine):
    Min. 8

Specification Test Results

About this item

 :
 :166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (1013 hPa)
Melting Pt:681 °C
Density:3.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-659-4
 :GHS08
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405