Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate, Certipur®, Supelco®
Oxidizing
Toxic
Corrosive
Health
Environment
Potassium dichromate, Certipur®, Supelco®
Supplier:  Merck
Certificates
Oxidizing
Toxic
Corrosive
Health
Environment
Potassium dichromate, Certipur®, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    130 g/l (20 °C)
  • Melting Point:
    398 °C
  • Molar Mass:
    294.19 g/mol
  • Bulk Density:
    1250 kg/m3
  • Boiling Point:
    500 °C (1013 hPa)
  • Density:
    2.69 g/cm3 (20 °C)
  • pH:
    4.0 (10 g/l, H2O)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Frequently Bought Together