ContactUsLinkComponent
Nalgene® Membrane Filters
Nalgene® Membrane Filters
   Sustainable Options Available
Certificates
undefined
Nalgene® Membrane Filters
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 513-2101
  • NALGDS0205-6045
  • 513-2152
  • 513-2151
  • 513-2052
  • 513-2102
  • Type:
  • 210
  • 205
  • 215
  • 215
  • 205
  • 210
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Pore size:
  • 0,20 µm
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,45 µm
  • Water flow rate:
  • >21 ml/min/cm² (1 bar)
  • >51 ml/min/cm² (1 bar)
  • >37 ml/min/cm² (1 bar)
  • >19 ml/min/cm² (1 bar)
  • >51 ml/min/cm² (1 bar)
  • >51 ml/min/cm² (1 bar)
  • Max. temperature:
  • 120 °C
  • 130 °C
  • 125 °C
  • 125 °C
  • 130 °C
  • 120 °C
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement