ContactUsLinkComponent
Spark-free MediLine laboratory freezers with ATEX certificate, LGUex and LGex
Spark-free MediLine laboratory freezers with ATEX certificate, LGUex and LGex
Supplier:  Liebherr
Certificates
undefined
Spark-free MediLine laboratory freezers with ATEX certificate, LGUex and LGex
Supplier:  Liebherr
Specifications
  • Catalog No:
  • 471-0483
  • Model:
  • LGUex 1500 MediLine
  • Cat. No.:
  • 471-0483
  • Display:
  • Digital
  • Defrost:
  • Manual
  • Alarm:
  • Audible and visual
  • Shelves/drawers:
  • 3 drawers, 1 basket
  • Door type:
  • 1 solid, reversible, self-closing door
  • Door lock:
  • Yes
  • Weight:
  • 42 kg
Specifications
Related Information
Product Information
Frequently Bought Together