
Sodium chloride, ULTREX®, Ultrapure, J.T.Baker®
Catalog # 4924-05
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
:GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477