Order Entry
Belgium
ContactUsLinkComponent
Lithium chloride ≥99%, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Lithium chloride ≥99%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Irritant
Lithium chloride ≥99%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99 %
  • Barium (Ba):
    max. 0.003 %
  • Calcium (Ca):
    max. 0.01 %
  • Iron (Fe):
    max. 0.001 %
  • Potassium (K):
    max. 0.01 %
  • Sodium (Na):
    max. 0.20 %
  • Heavy Metals (as Pb):
    max. 0.002 %
  • Insoluble Matter:
    max. 0.01 %
  • Loss on Drying:
    max. 1.0 %
  • Sulfate (SO4):
    max. 0.01 %
  • Titrable Base (meq/g):
    max. 0.008

Specification Test Results

Related Information

Frequently Bought Together