Order Entry
Belgium
ContactUsLinkComponent
 
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, Ultrapure Bioreagent, J.T.Baker®
Catalog # 4001-01
Supplier:  Avantor
CAS Number:  
undefined
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, Ultrapure Bioreagent, J.T.Baker®
Catalog # 4001-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 100.5 %
  • Assay (KCl) (by Ag titrn):
    99.0 - 100.5 %
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • Loss on Drying at 105°C:
    ≤1.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    5.4 - 8.6
  • Iodide (I):
    ≤0.002 %
  • Bromide (Br):
    ≤0.1 %
  • Barium (Ba):
    ≤0.001 %
  • Calcium, Magnesium, and R₂O₃ Precipitate:
    ≤0.005 %
  • Sodium (Na):
    ≤0.005 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Arsenic (As):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm
  • Trace Impurities - Nickel (Ni):
    ≤1.000 ppm

Specification Test Results

About this item

 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360