Order Entry
Belgium
ContactUsLinkComponent
Potassium chloride, BAKER, Laboratory Reagent, J.T.Baker®
Potassium chloride, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Potassium chloride, BAKER, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99 %
  • Acidity and Alkalinity:
    passes test
  • Iron (Fe):
    max. 0.005 %
  • Sodium (Na):
    max. 0.1 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Sulfate (SO4):
    max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together