
Cesium chloride ≥99.9995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:642 °C
Density:3.98 g/cm³ (20 °C)
EINECS:231-600-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.5 %
- Aluminium (Al):Max. 0.02 ppm
- Arsenic (As):Max. 0.005 ppm
- Barium (Ba):Max. 0.5 ppm
- Beryllium (Be):Max. 0.005 ppm
- Bismuth (Bi):Max. 0.05 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.02 ppm
- Cerium (Ce):Max. 0.01 ppm
- Cobalt (Co):Max. 0.005 ppm
- Chromium (Cr):Max. 0.01 ppm
- Copper (Cu):Max. 0.005 ppm
- Dysprosium (Dy):Max. 0.01 ppm
- Erbium (Er):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Iron (Fe):Max. 0.05 ppm
- Gallium (Ga):Max. 0.005 ppm
- Gadolinium (Gd):Max. 0.01 ppm
- Mercury (Hg):Max. 5 ppb
- Holmium (Ho):Max. 0.01 ppm
- Indium (In):Max. 0.005 ppm
- Potassium (K):Max. 1 ppm
- Lanthanum (La):Max. 0.01 ppm
- Lithium (Li):Max. 0.1 ppm
- Lutetium (Lu):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 1 ppm
- Neodymium (nd):Max. 0.01 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.01 ppm
- Praseodymium (Pr):Max. 0.01 ppm
- Rubidium (Rb):Max. 2 ppm
- Antimony (Sb):Max. 0.05 ppm
- Scandium (Sc):Max. 0.01 ppm
- Selenium (Se):Max. 0.05 ppm
- Samarium (Sm):Max. 0.01 ppm
- Strontium (Sr):Max. 0.1 ppm
- Terbium (Tb):Max. 0.01 ppm
- Thorium (Th):Max. 0.01 ppm
- Titanium (Ti):Max. 0.01 ppm
- Thallium (Tl):Max. 0.05 ppm
- Thulium (Tm):Max. 0.01 ppm
- Uran (U):Max. 0.01 ppm
- Vanadium (V):Max. 0.01 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Zinc (Zn):Max. 0.02 ppm
- Total N:Max. 10 ppm
- Total S (as SO₄):Max. 20 ppm
Specification Test Results
Frequently Bought Together