Order Entry
Belgium
ContactUsLinkComponent
Cesium chloride, Suprapur® for trace analysis, Supelco®
Health
Cesium chloride, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
Health
Cesium chloride, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1860 g/L (20 °C)
  • Melting Point:
    646 °C
  • Molar Mass:
    168.36 g/mol
  • Bulk Density:
    1800 kg/m³
  • Boiling Point:
    1382 °C
  • Density:
    3.97 g/cm³ (20 °C)
  • pH:
    7 - 9 (50 g/L, H₂O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together