Order Entry
Belgium
ContactUsLinkComponent
Cesium chloride ≥99.9%, Ultrapure Bioreagent for density gradient ultracentrifugation, J.T.Baker®
Cesium chloride ≥99.9%, Ultrapure Bioreagent for density gradient ultracentrifugation, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Cesium chloride ≥99.9%, Ultrapure Bioreagent for density gradient ultracentrifugation, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.9 %
  • Assay (as CsCl):
    ≥ 99.9 %
  • Absorbance of 50% w/v Aqueous Solution at 260 nm (1-cm path):
    ≤ 0.020
  • RNase Activity:
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • Protease Activity:
    Passes Test
  • ACS - Sulfate (SO₄):
    ≤ 5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 10.0 ppm
  • Trace Impurities - Barium (Ba):
    ≤ 5.0 ppm
  • Trace Impurities - Calcium (Ca):
    ≤ 10.0 ppm
  • Trace Impurities - Chromium (Cr):
    ≤ 5.0 ppm
  • Trace Impurities - Copper (Cu):
    ≤ 3.0 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 2.0 ppm
  • Trace Impurities - Lead (Pb):
    ≤ 5 ppm
  • Trace Impurities - Lithium (Li):
    ≤ 5.0 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤ 3.0 ppm
  • Trace Impurities - Manganese (Mn):
    ≤ 5.0 ppm
  • Trace Impurities - Potassium (K):
    = 5.0 ppm
  • Trace Impurities - Rubidium (Rb):
    ≤ 10.0 ppm
  • Trace Impurities - Strontium (Sr):
    ≤ 5.0 ppm

Specification Test Results

Related Information

Frequently Bought Together