Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate, anhydrous, EMSURE® ISO for analysis, Supelco®
Irritant
Sodium carbonate, anhydrous, EMSURE® ISO for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Irritant
Sodium carbonate, anhydrous, EMSURE® ISO for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    220 g/l (20 °C)
  • Melting Point:
    854 °C
  • Molar Mass:
    105.99 g/mol
  • Bulk Density:
    1100 kg/m3
  • Boiling Point:
    1600 °C (decomposition)
  • Density:
    2.53 g/cm3 (20 °C)
  • pH:
    11.5 (50 g/l, H2O, 25 °C)

Specification Test Results

Related Information

Frequently Bought Together