Order Entry
Belgium
ContactUsLinkComponent
Lithium carbonate ≥99%, GPR RECTAPUR®
Irritant
Lithium carbonate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Lithium carbonate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Insolubles:
    Max. 0.03 %
  • Iron (III) oxide:
    Max. 20 ppm
  • Water:
    Max. 0.5 %
  • Cl (Chloride):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 0.08 %
  • Ca (Calcium):
    Max. 0.12 %
  • K (Potassium):
    Max. 0.05 %
  • Na (Sodium):
    Max. 0.12 %

Specification Test Results

Frequently Bought Together