Order Entry
Belgium
ContactUsLinkComponent
Lithium carbonate, Suprapur® for trace analysis, Supelco®
Irritant
Lithium carbonate, Suprapur® for trace analysis, Supelco®
Catalog # 1.05676.0250
Supplier:  Merck
CAS Number:  
Irritant
Lithium carbonate, Suprapur® for trace analysis, Supelco®
Catalog # 1.05676.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    13 g/l (20 °C)
  • Melting Point:
    720 °C
  • Molar Mass:
    73.89 g/mol
  • Bulk Density:
    250 kg/m3
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.1 g/cm3 (20 °C)
  • pH:
    10 - 11 (5 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :73.89 g/mol
Merck Index:13,05549
Melting Pt:720 °C
Density:2.11 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:209-062-5
 :GHS07
Storage Temperature:Ambient
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084