


Potassium bromate ≥99.8%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7758-01-2
:
Formula:KBrO₃
MDL Number:MFCD00011359
:167 g/mol
Merck Index:12,07779
Melting Pt:350 °C
Density:3.218 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-829-8
ADR:5.1,II
:M-Clarity™ quality level = MQ300
UN:1484
:GHS03
:GHS06
:GHS08
Specification Test Results
- Assay (iodometric):≥99.8%
- Insoluble matter:≤ 0.005%
- pH value (5%;water;25°C):5.0-9.0
- Bromide (Br):≤ 0.02%
- Sulphate (SO₄):≤ 0.005%
- Total nitrogen (N):≤ 0.001%
- Heavy metals (as Pb):≤ 0.0005%
- Fe (Iron):≤ 0.0005%
- Na (Sodium):≤ 0.01%
Specification Test Results
Related Information
Frequently Bought Together