Sodium metaarsenite 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium dioxoarsenate
CAS Number:7784-46-5
:
Formula:NaAsO₂
MDL Number:MFCD00003472
:129.91 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:232-070-5
UN:1686
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.04975 - 0.05025
- Measurement uncertainty:+/- 0.00015
- Traceability:NIST SRM
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur® Merck Supelco® Titration goes Digital
Product Literature
msds_106277
SDS
ProductPage_106277
Product specification
Frequently Bought Together