Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Methylcyclohexane ≥99%, TECHNICAL

Flammable
Health
Irritant
Environment
Supplier:   VWR Chemicals
Methylcyclohexane ≥99%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Methylcyclohexane ≥99%, TECHNICAL

2 Options Available Below

Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:13,06074
  • Boiling Pt:101 °C (1013 hPa)
  • Melting Pt:–126 °C
  • Density:0.7693 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 99 %
  • Identification:
    Passes test
  • Water:
    Max. 50 ppm
Product Family Options

Product Information