Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cyclopentane 97%, extra pure

Flammable
Health
Irritant
Cyclopentane 97%, extra pure
Certificates
Cyclopentane 97%, extra pure

3 Options Available Below

Product Details & Documents
Synonyms:  Pentamethylene
  • CAS Number:287-92-3
  • Formula:C₅H₁₀
  • MDL Number:MFCD00001356
  • Molecular Weight:70.13 g/mol
  • Merck Index:13,02769
  • Boiling Pt:50 °C (760 mmHg)
  • Melting Pt:–94 °C
  • Density:0.75 g/cm³
  • Flash Pt:–7 °C
  • Storage Temperature:Ambient
  • EINECS:206-016-6
  • UN:1146
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4050 to 1.4070 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Residue after evaporation:
    ≤0.01 %
Product Family Options

Product Information