1-Heptanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis)
: TCI
:22767-50-6
:
:H₃C(CH₂)₅CH₂SO₃Na
:MFCD00007543
:202.25 g/mol
:300 °C
:Ambient
:245-210-5
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together