Order Entry
Belgium
ContactUsLinkComponent
 
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
 
undefined
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
 

 

  • Assay (GC):
    Min. 99.50 %
  • Water:
    Max. 0.0100 %
  • Non-volatile residue:
    Max. 0.0005 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Transmittance (230 nm):
    Min. 75.0 %
  • Transmittance (240 nm):
    Min. 80.0 %
  • Transmittance (250 nm):
    Min. 98.0 %

 

Frequently Bought Together