Order Entry
Belgium
ContactUsLinkComponent
2-Butoxyethanol ≥99%
2-Butoxyethanol ≥99%
Catalog # 537551-1L
CAS Number:  
undefined
2-Butoxyethanol ≥99%
Catalog # 537551-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.0 %
  • Density (20°C):
    0.9000 - 0.9040 g/ml
  • Water (Karl Fischer):
    Max. 0.10 %
  • Free acid (as CH₃COOH):
    Max. 0.010 %
  • APHA:
    Max. 10
  • Vapor Pressure:
    <1 mmHg ( 20 °C)

Specification Test Results

About this item

 :
 :118.18 g/mol
Merck Index:13,01558
Boiling Pt:168-172 °C (1.013 hPa)
Boiling Pt:168…172 °C (1.013 hPa)
Melting Pt:-74.8 °C
Melting Pt:–74.8 °C
Density:0.9 g/cm³ (20 °C)
Flash Pt:10.6 %(V)
Storage Temperature:Ambient
EINECS:203-905-0
ADR:6.1,III
 :GHS07
CAS Number:111-76-2
Formula:CH₃(CH₂)₃OCH₂CH₂OH
MDL Number:MFCD00002884