
Potassium hydroxide 0.5 M KOH (0.5N) volumetric solution, Fluka™
35116-1L
:
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
:
:56.11 g/mol
:1.023 g/cm³
:Ambient
:215-181-3
:1814
:8,II
:GHS05
:1310-58-3
:KOH
:MFCD00003553