Order Entry
Belgium
ContactUsLinkComponent
Cyclohexane ≥99%, GPR RECTAPUR®
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Substances coloured by H2SO4:
    Passes test
  • Boiling point:
    79 - 81 °C
  • Density (20/4):
    0.778 - 0.779
  • Free acidity:
    Max. 0.0002 meq/g
  • Solidification point:
    5.5 - 6.5 °C
  • Evaporation residue:
    Max. 50 ppm

Specification Test Results

Frequently Bought Together