Order Entry
Belgium
ContactUsLinkComponent
Cadmium(II) sulphate 8/3-hydrate 98.0-102.0%, Solarpur®, Sigma-Aldrich®
Cadmium(II) sulphate 8/3-hydrate 98.0-102.0%, Solarpur®, Sigma-Aldrich®
Catalog # 8.45500.1000
Supplier:  Merck
CAS Number:  
Cadmium(II) sulphate 8/3-hydrate 98.0-102.0%, Solarpur®, Sigma-Aldrich®
Catalog # 8.45500.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (complexometric, CdSO₄.8/3 H₂O):
    98.0 - 102.0 %
  • Identity Cd (ICP):
    passes test
  • Identity (SO₄):
    passes test
  • Insoluble matter:
    ≤ 0.005 %
  • pH-value (5 %; water):
    3.0 - 6.0
  • Chloride (Cl):
    ≤ 0.001 %
  • Ca (Calcium):
    ≤ 0.005 %
  • Cu (Copper):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.0005 %
  • K (Potassium):
    ≤ 0.01 %
  • Na (Sodium):
    ≤ 0.005 %
  • Pb (Lead):
    ≤ 0.002 %
  • Zn (Zinc):
    ≤ 0.002 %
  • Al (Aluminium):
    ≤ 50 ppm
  • Co (Cobalt):
    ≤ 50 ppm
  • Ni (Nickel):
    ≤ 50 ppm
  • Cr (Chromium):
    ≤ 50 ppm
  • Mn (Manganese):
    ≤ 50 ppm
  • As (Arsenic):
    ≤ 50 ppm
  • Sn (Tin):
    ≤ 50 ppm
  • Sb (Antimony):
    ≤ 50 ppm
  • Li (Lithium):
    ≤ 50 ppm

Specification Test Results

About this item

Synonyms: Cadmium sulphate, 8/3-hydrate, Cadmium sulphate octahydrate
 :
 :256.51 g/mol (anhy)
Merck Index:13,01627
Melting Pt:41 °C
Density:3.79 g/cm³ (20 °C)
EINECS:233-331-6
UN:2570
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :GHS09
CAS Number:7790-84-3
Formula:3CdSO₄·8H₂O
MDL Number:MFCD02253000