
Terpineol, mixture of isomers, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:8000-41-7
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00166983
:154.25 g/mol
:M-Clarity™ quality level = MQ200
EINECS:232-268-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Molar Mass:154.25 g/mol
- Boiling Point:214 - 217 °C
- Vapor Pressure:0.24 hPa (20 °C)
- Flash Point:91 °C
- Density:0.930 - 0.935 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together