
Brij® 35, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9002-92-0
:
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891
:1199.56 g/mol
:M-Clarity™ quality level = MQ200
EINECS:500-002-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(25 °C) soluble
- Melting Point:36 - 42 °C
- Molar Mass:582.8 g/mol
- Bulk Density:1050 kg/m3
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:149 °C
- Density:1.05 g/cm3 (20 °C)
- pH:5.5 - 7.0 (100 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together