Cyclohexane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–18 °C
:GHS02
:GHS08
:GHS07
:GHS09
Related Information
Certificates
Search by Lot NumberMERC1.02827.1000_TDS_EN ProductPage_102827 ProductPage_102827
Product specification
msds_102827
SDS
Frequently Bought Together