Order Entry
Belgium
ContactUsLinkComponent
Brij® 35 30% in aqueous solution
Brij® 35 30% in aqueous solution
Catalog # KRAF04214.3000
Supplier:  Bernd Kraft
CAS Number:  
Brij® 35 30% in aqueous solution
Catalog # KRAF04214.3000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :1199.56 g/mol
EINECS:500-002-6
CAS Number:9002-92-0
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891