Order Entry
Belgium
ContactUsLinkComponent
Brij® 35 10% in aqueous solution
Irritant
Brij® 35 10% in aqueous solution
Catalog # BMOL56256-1
CAS Number:  
Irritant
Brij® 35 10% in aqueous solution
Catalog # BMOL56256-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

About this item

 :
 :1199.56 g/mol
EINECS:500-002-6
 :GHS07
Storage Temperature:Ambient
CAS Number:9002-92-0
Formula:C₅₈H₁₁₈O₂₄
MDL Number:MFCD00080891