Order Entry
Belgium
ContactUsLinkComponent
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Certificates
undefined
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Vapor Density:
    2.9 (vs air)
  • Vapor Pressure:
    168.8 mmHg ( 37.7 °C)
  • Explosion Limit:
    9 %
  • Autoignition Temperature:
    500 °F

Specification Test Results

Related Information

Frequently Bought Together