



Cyclohexane
Catalog # EHERC11824500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814