Cyclohexane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9292-03
Supplier: Avantor
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.08
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.30
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤205
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.5 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (C₆H₁₂) (by GC, corrected for water):≥99.5 %
- Residue after Evaporation:≤2.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤0.01 %
Specification Test Results
About this item
:
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
:Packaged under nitrogen.
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814