Order Entry
Belgium
ContactUsLinkComponent
Bis(triphenylphosphine)palladium(II) chloride 98%
Irritant
Bis(triphenylphosphine)palladium(II) chloride 98%
Certificates
undefined
Irritant
Bis(triphenylphosphine)palladium(II) chloride 98%

Specification Test Results

  • Appearance:
    Yellow Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Assay:
    14.8 to 15.5 % (Pd)

Specification Test Results

Related Information

Frequently Bought Together