
Tris(triphenylphosphine)ruthenium(II) chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15529-49-4
:
Formula:C₅₄H₄₅Cl₂P₃Ru
MDL Number:MFCD00013077
:958.85 g/mol
Melting Pt:159 °C (dec.)
Storage Temperature:Ambient
EINECS:239-569-7
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Red-brown to black Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Carbon (C):66.0 to 69.0 %
Specification Test Results
Frequently Bought Together