Order Entry
Belgium
ContactUsLinkComponent
 
 
Bis(triphenylphosphine)palladium(II) chloride (15% Pd) for analysis
 
undefined
 
Bis(triphenylphosphine)palladium(II) chloride (15% Pd) for analysis

 

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    14.6 to 15.8 % (Pd)
  • Calcium (Ca):
    ≤15 ppm
  • Copper (Cu):
    ≤10 ppm
  • Gold (Au):
    ≤10 ppm
  • Iridium (Ir):
    ≤10 ppm
  • Iron (Fe):
    ≤5 ppm
  • Lead (Pb):
    ≤5 ppm
  • Magnesium (Mg):
    ≤5 ppm
  • Platinum (Pt):
    ≤10 ppm
  • Rhodium (Rh):
    ≤10 ppm
  • Ruthenium (Ru):
    ≤10 ppm
  • Silicon (Si):
    ≤10 ppm
  • Silver (Ag):
    ≤10 ppm

 

 

Frequently Bought Together