Order Entry
Belgium
ContactUsLinkComponent
 
Cyclohexane ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
  9258-03
 :  Avantor
 :  
undefined
Cyclohexane ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
  9258-03
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 10
  • Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤10
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤10
  • Assay (C₆H₁₂) (by GC, corrected for water):
    ≥99.0 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤2.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤100 ppm

 

 

 :
 :84.16 g/mol
 :13,02752
 :81 °C (1013 hPa)
 :7 °C
 :0.7785 g/cm³ (20 °C)
 :–18 °C
 :Ambient
 :203-806-2
 :1145
 :3,II
 :Packaged under inert gas.
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :110-82-7
 :C₆H₁₂
 :MFCD00003814