Order Entry
Belgium
ContactUsLinkComponent
 
Organic reference standard, Diisodecyl phthalate 100 µg/ml in methanol
Catalog # ACCUALR-101S
Supplier:  AccuStandard
CAS Number:  
Organic reference standard, Diisodecyl phthalate 100 µg/ml in methanol
Catalog # ACCUALR-101S
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    26761-40-0
  • Description:
    Diisodecyl phthalate 100 µg/ml in methanol
  • Formula:
    C₂₈H₄₆O₄
  • Description:
    Diisodecyl phthalate

Specifications

About this item