ContactUsLinkComponent
Cyclohexane, BAKER, Laboratory Reagent, J.T.Baker®
 :  Avantor
undefined
Cyclohexane, BAKER, Laboratory Reagent, J.T.Baker®
 :  Avantor
  • For Laboratory,Research,or Manufacturing Use:
    min. 99 %
  • Residue after Evaporation:
    max. 0.005 %
  • Water (H2O):
    max. 0.02 %
Frequently Bought Together