Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cyclohexane 99+%, pure

Cyclohexane 99+%, pure

Irritant
Health
Environment
Flammable
Cyclohexane 99+%, pure
Certificates
Cyclohexane 99+%, pure

5 Options Available Below

Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (K.F.)
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.005 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 72,20 €
    72,20 €
    1 * 1 L
       72,20 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 148,00 €
    148,00 €
    1 * 2,5 L
       148,00 €

  • Plastic drum
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 252,00 €
    252,00 €
    1 * 5 L
       252,00 €

  • Plastic drum
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 404,00 €
    404,00 €
    1 * 10 L
       404,00 €

  • Plastic drum
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 960,00 €
    960,00 €
    1 * 25 L
       960,00 €